About this item
PHCbi's large capacity pharmaceutical refrigerators deliver stable, reliable temperature control for clinical, pharmaceutical, biomedical storage and processing at uniform temperatures. Easy to use features and design and precise temperature control with fast recovery following door openings, assures safe storage during frequent access and high ambient conditions.
- Temperature stability
- Wide temperature range
- Speedy and powerful refrigeration
- Cycle defrost
- Filterless condenser cuts down on cleaning maintenance
- Standard alarm and safety features
- HFC and HFO combined (CFC-free) refrigerant and CFC-free insulation
- Top-mounted air circulation blower
These pharmaceutical refrigerators are configured to provide close temperature control for both storage and process applications. A powerful refrigeration system delivers fast temperature pull-down, recovery and tolerance for high ambient temperatures. SNAP compliant refrigerants and energy efficiency helps improve LEED scores.
The MPR Series offers two choices in inventory control. The MPR-1412-PA includes 8 open-wire shelves strong enough to hold bulk loads. The MPR1412R-PA is equipped with 8 heavyduty, fully-extendable painted steel pull-out drawers for more convenient access to stored product such as large bottles or reagent kits.
Specifications
- Catalog No:
- CA76355-510
- CA76355-512
- Cat. No.:
- 76355-510
- 76355-512
- Display:
- LED
- LED
- Electrical:
- 115 V
- 115 V
- Weight:
- 248 kg (547 lbs.)
- 287 kg (637 lbs.)