PIG® Green Ham-O® Absorbent Mat Roll, New Pig
Catalog #: 76378-318
Supplier: New Pig
Specifications
- For Use With:Oils, coolants, solvents, water – Universal
- Description:PIG® Green Ham-O® Absorbent mat roll
- Cat. no.:76378-318
- Packaging:1 roll per bag
- Absorbency:47.4 gal./box
- Dimensions:32" × 150' roll
Specifications
About this item
Ham-O layer camouflages liquids and grime while it absorbs; tear off only what you need.
- Absorbed leaks and drips blend right in to camouflage top layer so mat appears clean
- Scuff-resistant hamouflage layer stands up to foot traffic in aisles and walkways; resists rips, tears and abrasions from tools when used as a liner
- Use anywhere – line workbenches, counters and toolboxes, catch drips and overspray around machinery, clean up oil and grease in service pits and more
- Easy-tear perforations let you tear off small or large pieces for less waste
- Absorbs most common industrial liquids - oils, coolants, solvents, water and more
- Can be wrung out and incinerated after use to reduce waste or for fuels blending
- Mats take on the properties of absorbed liquids - always take proper precautions when handling saturated absorbents
PIG-Printed Camouflage Mats are a fun way to absorb liquid and hide grime; Ham-O mats keep your workplace clean and your employees safe and happy.
No job is too big for these wide heavyweight rolls; the mat weight is ideal for absorbing leaks, drips or overspray in walkways and aisles and for soaking up big spills in large areas.
PIG Mat lasts 2× longer than competitive mats and won't rip, tear or fray even when saturated; liquids absorb quickly for faster, easier cleanups and safer workplaces.
Customers Who Bought This Also Bought