Corepoint Scientific™ Undercounter Controlled Room Temperature (CRT) Chambers, Horizon Scientific
Supplier: Horizon Scientific
Certificates
About this item
These CRT cabinets provide controlled room temperature environments for storing sensitive substances that require precise temperature management.
- Integrated temperature alarms
- Keyed door locks
- Forced draft air circulation
- Probe access
- Precise microprocessor temperature control
- Natural hydrocarbon refrigerants
These specialized cabinets are ideal for scientific and medical applications. Built-in units are designed with front-facing ventilation systems, allowing easy installation into a bench or countertop. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Ordering information: All units have a two-year parts and labor warranty, plus an additional three-year compressor parts warranty.
Specifications
- Catalog No:
- 76403-506
- 76403-508
- Ext. W×D×H:
- 451W×578D×781H mm (17¾×22¾×30¾")
- 451W×578D×781H mm (17¾×22¾×30¾")
- Environmental certification:
- Natural refrigerants
- Natural refrigerants
Specifications
Related Information
Exterior Cutsheet 76403-506 CRTPR031WWW0FB.pdf Exterior Cutsheet 76403-508 CRTPR031WWWBLH.pdf Interior Cutsheet 76403-506 CRTPR031WWW-0FB.pdf Interior Cutsheet 76403-508 CRTPR031WWWBLH.pdf
Technical Drawing
TDS Undercounter Controlled Room Temperature Cabinet Left Hinged TDS Undercounter Controlled Room Temperature Cabinet, Built-In 2.5 Cu. Ft. Capacity.pdf TDS Undercounter Controlled Room Temperature Cabinet, Left Hinged Swing Door.pdf TDS Undercounter Controlled Room Temperature Cabinet, Right Hinged Swing Door.pdf
Datasheet
Customers Who Bought This Also Bought