Clinisorb Non-Woven Sponges, DUKAL™ Corporation
Supplier: Dukal
Certificates
About this item
Multi-purpose sponge dressings are ideal for general wound care and applying ointments.
- Rayon/poly blend construction
- Highly absorbent
- Virtually non-linting
- No natural rubber latex
Available in non sterile and sterile formats.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.