ContactUsLinkComponent
Heratherm™ General Protocol Microbiological Incubators, Thermo Scientific
Heratherm™ General Protocol Microbiological Incubators, Thermo Scientific
Certificates
Heratherm™ General Protocol Microbiological Incubators, Thermo Scientific
Heratherm™ General Protocol Microbiological Incubators, Thermo Scientific
Certificates
Specifications
  • Catalog No:
  • 76411-573
  • 76411-577
  • 76411-628
  • 76411-632
  • 76411-636
  • Temperature range:
  • Ambient +5…75 °C
  • Ambient +5…75 °C
  • Ambient +5…75 °C
  • Ambient +5…75 °C
  • Ambient +5…75 °C
  • Electrical:
  • 120 V, 60 Hz
  • 120 V, 60 Hz
  • 120 V, 60 Hz
  • 120 V, 60 Hz
  • 120 V, 60 Hz