Specifications
- Description:ClonePix™ 2 mammalian colony picker
- Camera:High resolution CMOS camera: <90 sec image acquisition in standard mode, Standard: 3.6 micron resolution
- Cat. no.:76404-534
- Dimensions:1010×900×1490 mm
- Weight:350 kg
Specifications
About this item
Redefine clone screening and selection: transform your cell line development workflow.
- Screen more clones in less time
- Accurate, automatic colony picking avoids errors associated with limiting dilution
- Excellent image quality allows for screening of stable, high-producing clones
- Increased productivity of a cell line development workflow
- Add FL and white light imaging
- > 200 clones per hour
The ClonePix™ 2 Mammalian Colony Picker is a fully automated system for the selection of high-value clones used in antibody discovery and cell line development. Hybridoma, CHO cells, and other cell types are imaged and selected based on user-defined parameters. Plate handling, barcode reading, and picking are all fully integrated. All data, including images, are saved for downstream analysis. The picker increases the probability of finding optimally produced cell lines and significantly reduces time and labor.
Source plate - PetriWell-6 plate, PetriWell-1 plate, Greiner 6-well plate, Nunc 6-well plate, Nunc OmniTray
Destination Plate - PetriWell-96 plate, Costar 96-well plate, Greiner 96-well plate, Nunc 96-well plate, Falcon 96-well plate
Trans-illumination for imaging low contrast colonies such as adherent mono layers or small colonies in suspension, Epi-illumination for imaging colonies as they are collected.
Uses 5 excitation /emission filter pairs, Fluorescence Filter Set Excitation 470nm, Emission 535nm (FITC) Fluorescence Filter Set Excitation 440nm, Emission 505nm (CFP) Fluorescence Filter Set Excitation 500nm, Emission 550nm (YFP) Fluorescence Filter Set Excitation 530nm, Emission 590nm (PE-R, Rhodamine, DS Red) Fluorescence Filter Set Excitation 623nm, Emission 700nm (Cy5)