Ryton® Scott Spray Chamber for Optima 3000 DV/3000 SCR/3×00 RL, PerkinElmer
Catalog #: CA97013-782
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Specifications
- Description:Ryton® scott spray chamber
- Cat. no.:CA97013-782
- Supplier No.:N0690640
Specifications
About this item
The Ryton® scott spray chamber is designed for use with Optima™ 3000, 3000 SCR, and 3×00 RL ICP-OES series instruments.
- Chamber is made from corrosion-resistant Ryton®
- Inert to most mineral acids including hydrofluoric acid, aqua regia, and all organic solvents normally used in ICP analyses
- Scott-type design minimizes pulsations from the peristaltic pump
The type II, two-piece design ensures leak-free performance. The removable inner tube allows the chamber to be easily disassembled for cleaning.
Customers Who Bought This Also Bought