Cesium (Cs) System 2 Electrodeless Discharge Lamp, PerkinElmer
Catalog #: CA97013-294
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Specifications
- Description:System 2 electrodeless discharge lamp
- Element:Cesium
- Supplier No.:N3050620
Specifications
About this item
System 2 Electrodeless Discharge Lamps (EDLs) provide the optimized spectral output needed to get the maximum performance from PerkinElmer® Atomic Absorption (AA) spectrometers.
- System 2 Electrodeless Discharge Lamp for the detection of elemental Cesium (Cs)
- PinAAcle™ instruments require the purchase of an EDL driver assembly for each of the lamp positions to be used
Electrodeless Discharge Lamps for greater brightness (EDLs) are typically much brighter and, in some cases, provide better sensitivity than comparable HCLs. EDLs are preferred for certain volatile elements. They offer better precision and lower detection limits for analyses that are “noisy” due to weak hollow cathode emission. Electrodeless Discharge Lamps (EDLs) are designed to be physically interchangeable with Intensitron Hollow Cathode Lamps. Their power supplies are built into all the PinAAcle™ series of atomic absorption spectrometers.
Customers Who Bought This Also Bought