Pen Clips, PerkinElmer
Catalog #: CA97013-736
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Specifications
- Description:Pen clips
- Cat. no.:CA97013-736
- Supplier No.:L4071029
Specifications
About this item
Beryl Copper pen clips for use with stainless steel ATD tubes.
Included in the Workplace Air Monitoring Industrial Hygiene Application Kit.
Customers Who Bought This Also Bought