Demountable Quartz Torch, 0-Slot for Optima™ 3000 SCX/3x00 XL, PerkinElmer
Catalog #: CA97012-916
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Specifications
- Description:0-Slot demountable quartz torch
- Cat. no.:CA97012-916
- For Use With:Optima™ 3000 SCX and 3x00 XL ICP-OES
- Supplier No.:N0695379
Specifications
About this item
This quartz torch is designed for use with Optima™ 3000 SCX and 3x00 XL ICP-OES series instruments.
This demountable 0-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow. It is recommended for use with organic solvents.
Customers Who Bought This Also Bought