Demountable Quartz Torch, 1-Slot for Optima 3x00 DV, PerkinElmer
Catalog #: CA97012-918
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Specifications
- Description:1-slot demountable quartz torch
- Cat. no.:CA97012-918
- For Use With:Optima™ 3x00 DV ICP-OES
- Supplier No.:N0691662
Specifications
About this item
This 1-slot quartz torch is designed for use with Optima™ 3x00 DV ICP-OES series instruments.
The demountable 1-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow, and is the standard torch for aqueous solutions.
Customers Who Bought This Also Bought