Complete Quartz Torch O-Ring Kit for Optima™ 2x00/4x00/5x00/7x00 DV Instruments, PerkinElmer
Catalog #: CA97012-772
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Specifications
- Description:Complete quartz torch O-ring kit
- Cat. no.:CA97012-772
- Supplier No.:N0770437
Specifications
About this item
This kit includes various torch O-rings used with Optima™ 2x00 DV, 4x00 DV, 5x00 DV, and 7x00 DV ICP-OES series instruments.
Delivery: This kit includes: (1) 29.74 mm Int.Ø O-Ring, (2) 20.3 mm Int.Ø O-Rings, (1) 28.17 mm Int.Ø O-Ring, (1) 26.57 mm Int.Ø O-Ring, and (1) 20.3 mm Int.Ø O-Ring.
Customers Who Bought This Also Bought