Specifications
- Description:Calcium (Ca) - Zinc (Zn) Lumina™ hollow cathode lamp
- Cat. no.:CA97013-290
- Supplier No.:N3050203
Specifications
About this item
Multi-Element Lumina™ hollow cathode lamp (HCL) for the detection of elemental Calcium (Ca) and Zinc (Zn). Series N30502XX Lumina™ 50 mm (2") diameter multi-element lamps are designed to be used with PinAAcle™ and AAnalyst™ Atomic Absorption spectrometer series of instruments.
Please note that multi-element lamps are not designed to be used in a furnace system.
Lamp is designed for use with and tested on spectrometers to assure compatibility and the highest performance. Offers a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ hollow cathode lamp (HCL), produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume - and the longer the lamp lifetime.