Specifications
- Description:Rhodium (Rh) Lumina™ hollow cathode lamp
- Cat. no.:CA97013-370
- Supplier No.:N3050166
Specifications
About this item
Single Element Lumina™ hollow cathode lamp (HCL) for the detection of elemental Rhodium (Rh). Series N30501XX Lumina™ 50 mm (2") diameter single-element lamps are designed to be used with our PinAAcle™ and AAnalyst™ atomic absorption spectrometer series of instruments.
Lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. Offers a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ hollow cathode lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume - and the longer the lamp lifetime.
Note: Lumina™ hollow cathode lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable.