MSA Mannitol Salt Agar Plates
Catalog #: 470192-610
Supplier: IPM SCIENTIFIC, INC.
CAS Number:
Specifications
- Packaging:Pack of 10
- Description:MSA plates
- Supplier No.:470192-610
Specifications
About this item
Prepared media plates.
- Microbiology
We recommend purchasing prepared media plates no more than two weeks before use for best results, as these items are highly perishable.
Customers Who Bought This Also Bought