Scalpel handles, Excelta Corp®
Supplier: Avantor
Certificates
About this item
These scalpel handles and blades with the exception of the 181-22, are made of AISI 440 stainless steel and are cleanroom safe.
Customers Who Bought This Also Bought
These scalpel handles and blades with the exception of the 181-22, are made of AISI 440 stainless steel and are cleanroom safe.