Specifications
- Description:StarSense Explorer DX 130AZ
- Cat. no.:470324-856
Specifications
About this item
StarSense Explorer is ideal for beginners thanks to the app’s user-friendly interface and detailed tutorials.
- Tour the night sky with your smartphone and the DX 130AZ telescope
- Lock targets for eyepiece viewing with in-app bullseye and slow-motion manual controls
- 130 mm (5-inch) Newtonian reflector with highly reflective optical coatings for excellent viewing
- Includes 10 mm and 25 mm eyepieces, smartphone dock, red dot finderscope, and full-height tripod
Leave complicated star charts, imprecise planetarium apps, and computerized mounts behind. With StarSense Explorer, locating objects has never been easier, faster, or more accurate. Within minutes of setting up the telescope, you’ll be navigating the sky with confidence. Simply place your phone in the unique StarSense dock and launch the StarSense Explorer app. After aligning your phone to the telescope’s optics (a quick, 2-minute procedure), StarSense Explorer generates a list of celestial objects currently visible. Make your selection and arrows appear onscreen, guiding you as you to move the telescope. When the object is ready to view, the bullseye turns green. StarSense Explorer works with most modern smartphones, including iPhone 6 and up and most devices running Android 7.1.2 or later manufactured since 2016.
StarSense Explorer uses patent-pending sky recognition technology and your smartphone to determine exactly where the telescope is pointed in the night sky. A Lost in Space Algorithm (LISA), like the ones satellites use in orbit to correctly reorient themselves, helps the app match star patterns it detects overhead to its internal database. While other astronomy apps may claim that they can help you find objects, they rely exclusively on the phone’s gyros and accelerometers, which aren’t as accurate as LISA technology. No other app can accurately tell you when your target is visible in the eyepiece.
High Quality Altazimuth Mount with Slow Motion Control:
An ultra-stable altazimuth mount provides a sturdy foundation for StarSense Explorer DX. Slip clutches and geared slow motion controls in both axes help you move the telescope smoothly and zero in your target. As celestial objects appear to drift across the night sky, you’ll be able to follow them with just a turn of the knobs. It’s all anchored by an adjustable, full-height tripod.
Dazzling Views with High Quality Optics:
With a large 130mm (5”) objective lens, this telescope has enough light gathering ability to bring out impressive detail in celestial objects. You can expect sharp, bright views of Jupiter’s four Galilean moons, its cloud bands and Great Red Spot, the rings of Saturn, the trapezium in the Orion Nebula, and beautiful Pleaides Open Star Cluster. Highly reflective coatings on both the primary and secondary mirror enhance the views even more.
Perfect for the city or dark sky sites. Even if you live in a light polluted city location, StarSense Explorer is advanced enough to be able to pick out Jupiter, Saturn, Venus, the Orion Nebula, double stars, and a few more of the most famous celestial objects. But if you can take the telescope to an even slightly darker location, more objects will become visible. With this 5” Newtonian and relatively dark skies, the Andromeda Galaxy, Hercules Open Star Cluster, and so many more are easily within your reach. The entire telescope kit weighs just 18 pounds, so it’s perfectly portable and easy to bring on your next camping trip or to a remote observing site.
Everything you need to observe immediately. When you unbox your new StarSense Explorer DX, you’ll find: 130mm Newtonian reflector optical tube, StarSense dock for your smartphone, Low powered (25mm) and high powered (10mm) eyepiece, StarPointer red dot finderscope, perfect for using the telescope during the day or without the StarSense Explorer app, Altazimuth mount with slow motion controls, Full-height tripod with an accessory tray to keep you organize.