Reaction Vessel Lid for CG-1968-W Style Reactors, Chemglass
Catalog #: 76333-376
Supplier: Chemglass
Specifications
- Description:Reaction vessel Lid complete, stainless steel, 400 mm, 7-neck, Large addition port
- Cat. no.:76333-376
Specifications
About this item
Reaction vessel lid is constructed of ¹/₄" thick #316 S.S. with a ¹/₈" thick PTFE gasket underneath. The only material exposed to the reaction is PTFE.
- The flat design reduces the height over the standard glass reaction vessel lids
Lid has seven openings that accept 45/50 outer PTFE inserts. One neck is a bulk addition port is 80 mm reaction flange with o-ring groove. Inserts are secured to the lid using glass filed PTFE nuts. Lid can be used on any of our CG-1968-W style reactors.
Customers Who Bought This Also Bought