Specifications
- Description:Lumina™ Hollow Cathode Lamp
- Element:Lithium
- Supplier No.:N3050142
Specifications
About this item
Single-element Lumina™ Hollow Cathode Lamp (HCL) for the detection of elemental Lithium (Li). Series N30501XX Lumina™ 2" (50 mm) diameter single-element lamps are designed to be used with PerkinElmer PinAAcle™ and AAnalyst™ Atomic Absorption (AA) spectrometer series of instruments.
- Lumina™ Hollow Cathode Lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable
Every genuine PerkinElmer lamp is designed for use with and tested on spectrometers to assure compatibility and high level performance. PerkinElmer offers a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ Hollow Cathode Lamp (HCL), they are produced with larger internal volume, providing a greater supply of fill gas at optimum pressure. The larger the lamp, the greater the inert gas volume, and the longer the lamp lifetime.