Superband UV/VIS/NIR Photometric Accuracy and Linearity Kit, FireflySci
Catalog #: 76016-848
Supplier: FIREFLYSCI, INC.
CAS Number:
Specifications
- Description:Superband UV/VIS/NIR photometric accuracy and linearity kit (10%T, 30%T, 90%T)
- Cat. no.:76016-848
- Tolerance:±0.0022 au
Specifications
About this item
Kit consists of three filters and a blank for validating photometric accuracy and linearity in the range of 200 to 3000 nm.
- Photometric accuracy in the UV and VIS range
- Optical densities: 0.04 au (90%), 0.5 au (30%), 1.0 au (10%)
- Wavelength range: 200 to 3000 nm
- External dimensions of individual filters: 12.5×12.5×45mm
Delivery: This kits includes the following filters: SPB-0.04 (90%T), SPB-0.5 (30%T), SPB-1.0 (10%T), A30 blank holder, and case P4.
Customers Who Bought This Also Bought