ContactUsLinkComponent
Micro Drop Spectrophotometer Conversion Kit, Micro Focus Cell (1.9 µl), FireflySci
Micro Drop Spectrophotometer Conversion Kit, Micro Focus Cell (1.9 µl), FireflySci
Catalog #: 76016-870
Supplier:  FIREFLYSCI, INC.
Micro Focus Cell (1.9ul)- Micro Drop Spectrophotometer Conversion Kit
Micro Drop Spectrophotometer Conversion Kit, Micro Focus Cell (1.9 µl), FireflySci
Catalog #: 76016-870
Supplier:  FIREFLYSCI, INC.
Specifications
  • Capacity:
    1.9 µl
  • Description:
    Micro drop spectrophotometer conversion kit
  • Cat. no.:
    76016-870
Specifications
About this item
Transform a current spectrophotometer into a nano-sized spectrophotometer with the Micro focus cell conversion kit.
  • Ultra-micro low volume cell with a 1.9 µl capacity
  • Convert any spectrophotometer into a high end drop based machine
  • Leak-proof design
Applications include DNA measurement, RNA measurement, DNA quantification, nucleic acid analysis, protein concentration, and all small volume applications (0.3 to 1.9 µl). Quickly load and empty a sample into the cell without even removing it from the machine. This results in consistent alignment through multiple tests.
Ordering information: Each kit includes a 12.5×12.5 micro focus adapter that fits into any spectrophotometer, a UV Quartz micro tray to inject a sample into, spacers to adjust the micro focus adapter to the proper Z-Dimension in the spectrophotometer, cleaning swabs, and instructions.
Customers Who Bought This Also Bought