About this item
Tamper-evident cryogenic tape for secure sealing of various cryogenic containers for storage in ultra-low temperature freezers and liquid nitrogen, and transport on dry ice. The cryo tape will tear-up upon any attempt to remove it, indicating whether the contents have been compromised.
- Temperature range: −196 to +110 °C (−321 to +230 °F)
- Tamper-evident seal ensures any attempt to open the container will be noticed
- Able to withstand immersion in liquid nitrogen and deep-freeze storage conditions
- Continuous tape format allows use of as much or little as necessary
- Available in assorted colors for easy identification and color-coding
Tamper-evident cryogenic tape for sealing ultra-low temperature freezer and cryogenic containers such as cryo vials, cryo/freezer boxes, cryo cassettes, and tubes. The continuous tape will tear-up upon any attempt to remove it from most surfaces, indicating whether the contents have been compromised. Durable and waterproof, the tamper-evident tape is available in a range of colors for easy identification. Writable using permanent cryogenic markers, the cryo tape is suitable for long-term storage in liquid nitrogen (−196 °C), laboratory freezers (−80 °C), and for transportation on dry ice.