Cryo-OmniTAG™, Transparent Cryogenic and Autoclave-Resistant Thermal-Transfer Labels, GA International
Supplier: GA INTERNATIONAL INC
Certificates
About this item
Transparent thermoplastic labels for a wide range of applications.
- Labels withstand long-term storage in cryogenic environments (–196 °C)
- Labels withstand sterilization by steam autoclaves, dry heat, and gamma irradiation
- Transparent, allowing constant view of container’s volume or surface
- Compatible with all major thermal-transfer barcode printers
From liquid nitrogen and freezer storage to autoclaving and dry-heat sterilization, Cryo-OmniTAG™ labels are suitable for the identification of cryovials, tubes, freezer boxes, glass bottles, and other surfaces. Thermal-transfer printable. Waterproof and weatherproof.
Customers Who Bought This Also Bought