Cryo-Marker™ Dual Point Cryogenic Water-Resistant Markers, GA International
Supplier: Avantor
Certificates
About this item
Dual point high-quality permanent ink marker.
- Excellent performance on plastic, glass, metal, cloth, and other surfaces
- Compatible with Lab-TAG™ Labels from GA International
- Easy to handle for applications in bio-medical laboratories
- Extra-Fine (0.3 mm) and fine (0.7 mm) tips; 2 pens in 1
- Can write on Teflon surfaces
- Low odor
Durable, heavy-duty, long lasting markers withstand rough treatment and provide delicate touch on various materials. Permanent and waterproof, all colors are equally permanent and lasting.
Customers Who Bought This Also Bought