Avantor® ACE® C18-300, HPLC Columns, 10 µm
Supplier: HICHROM LIMITED
Certificates
About this item
Avantor® ACE® C18-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative. These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
- Designed for peptide and protein separations
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Ideal for a wide range of analytes differing in hydrophobicity
- Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
These columns provide excellent peak shape even at extremely low levels of additives such as TFA, making them ideal for LC/MS applications.
Ordering information: Method validation kits contain three different batches of the same phase - available on request.
Specifications
- Catalog No:
- 76391-214
- 76391-206
- 76391-208
- 76391-232
- 76391-190
- 76391-192
- 76391-240
- 76391-238
- 76391-236
- Carbon load:
- 9.0%
- 9.0%
- 9.0%
- 9.0%
- 9.0%
- 9.0%
- 9.0%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
Specifications
Related Information
Avantor® ACE® HPLC and UHPLC Columns
Product Literature
TDS Avantor ACE® HPLC and UHPLC Columns
Product specification
Customers Who Bought This Also Bought