Avantor® Partisil PAC, HPLC Columns
Supplier: HICHROM LIMITED
Certificates
About this item
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
- Easy to use
- Irregular porous silica
- Stainless steel column hardware
Avantor® Partisil® PAC is a polar amino cyano bonded phase with secondary amine groups for good thermal and chemical stability. Fast equilibration enables multiple separation mechanisms, including adsorption, reversed-phase and weak anion-exchange, to be used. The phase is particularly suitable for carbohydrate separations.
Caution: * Use with freestanding holder HI-161 and column coupler HI-081, please see 'HPLC Guard Column Holders and Accessories'.
Customers Who Bought This Also Bought