Avantor® Partisil SAX, HPLC Columns
Supplier: HICHROM LIMITED
Certificates
About this item
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
- Easy to use
- Irregular porous silica
- High efficiency
- Stainless steel column hardware
Avantor® Partisil® SAX is a strong anion exchange phase based on quaternary ammonium groups and is stable over the pH range 1.5 to 7.5. At intermediate pH, use of a Solvecon eluent conditioning column is recommended to further enhance column lifetime. Suitable for use with nucleic acids, organic acids and inorganic anions.
Specifications
- Catalog No:
- 76401-970
- 76402-210
- 76402-216
- 76401-966
- 76401-956
- 76402-196
- 76401-960
- 76402-202
- 76402-200
- 76401-958
- 76401-950
- 76402-188
- 76401-946
- 76401-944
- 76402-218
- 76402-222
- Carbon load:
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- pH range:
- 1.5 - 7.5
- 1.5 - 7.5
- 1.5 - 7.5
- 1.5 - 7.5
- 1.5 - 7.5
- 1.5 - 7.5
- 1.5 - 7.5
- 1.5 - 7.5
- 1.5 - 7.5
- 1.5 - 7.5
- 1.5 - 7.5
- 1.5 - 7.5
- 1.5 - 7.5
- 1.5 - 7.5
- 1.5 - 7.5
- 1.5 - 7.5