Avantor® Partisil ODS2, HPLC Columns, 10 µm
Supplier: HICHROM LIMITED
Certificates
About this item
Avantor® Partisil® is one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity for preparative work.
- Irregular porous silica
- Convenient and easy to use
- Hand tightened fittings
- Increased column lifetime
- Stainless steel column hardware
Caution: Use with freestanding holder HI-161 and column coupler HI-081, please see 'HPLC Guard Column Holders and Accessories'.
Customers Who Bought This Also Bought