Avantor® Partisil ODS, HPLC Columns, 10 µm
Supplier: HICHROM LIMITED
Certificates
About this item
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity for preparative work.
- Irregular porous silica
- Convenient and easy to use: Hand tightened fittings
- Increased column lifetime
- Stainless steel column hardware
Low carbon load C18 phase, with intentionally unbonded surface silanols adding to the selectivity.
Caution: *Use with freestanding holder HI-161 and column coupler HI-081, please see "HPLC Guard Column Holders and Accessories.
Customers Who Bought This Also Bought