Avantor® Hichrom, Pliers for Stainless Steel Tubing
Catalog #: 76399-196
Supplier: HICHROM LIMITED
Specifications
- Description:Stainless steel tubing cutter pliers
- Cat. no.:76399-196
Specifications
About this item
These pliers are designed for cutting ¹/₁₆" stainless steel tubing and are especially suitable for tubing of 0.010" and 0.020" ID.
- Leaves smooth and uniform cuts
- De-burring may be necessary to achieve an optimum finish
Customers Who Bought This Also Bought