Avantor® Hichrom, ValvTool
Catalog #: 76399-202
Supplier: HICHROM LIMITED
Specifications
- Description:ValvTool
- Cat. no.:76399-202
Specifications
About this item
This Valvtool is a uniquely designed slotted wrench which provides easy access to many hard-to-reach areas.
It enables fittings, sample loops and most other ¹/₄" stainless steel and PEEK HPLC fittings to be more easily tightened or loosened.
Customers Who Bought This Also Bought