Avantor® Hichrom Last Drop™, Inlet Filters
Supplier: HICHROM LIMITED
Certificates
About this item
The Last Drop™ eluent filter utilises a flat element, which sits parallel to the bottom of the reservoir, allowing the filter to draw all but the last 2% of the eluent without drawing air into the system.
- Reduces solvent wastes
- Used for sensitive biochromatography applications
The filter consists of a 316 stainless steel or PTFE filter element in an inert housing.
Delivery: Every filter is supplied with a universal PEEK tubing adaptor.
Customers Who Bought This Also Bought