Avantor® Alltima HP C18, HPLC Columns
Supplier: HICHROM LIMITED
Certificates
About this item
Avantor® Alltima HP columns offer a range of different phase chemistries based on high purity silica. The Avantor® Alltima HP product family combines the selectivity and performance needed to overcome the most challenging separation needs.
- Excellent column stability
- Low to no detectable column bleed
- pH stability from 1 to 10
- Classic reversed-phase retention and selectivity
- Multiple selectivity options
The low column bleed makes these columns ideal for microbore applications.
Column hardware: Stainless steel
Specifications
- Catalog No:
- 75797-330
- 75797-328
- 75797-372
- 75797-334
- 75797-378
- 75798-604
- 75797-332
- 75797-376
- 75797-338
- 75797-382
- 75797-336
- 75797-340
- 75797-384
- 75830-814
- 75797-386
- 75830-816
- 75797-348
- 75798-606
- 75797-324
- 75797-322
- 75797-352
- 75797-350
- 75797-356
- 75797-354
- 75797-360
- 75797-320
- 75797-364
- 75797-362
- 75798-592
- 75798-594
- 76398-378
- 75798-600
- 75798-602
- 75797-326
- 75797-370
- 75798-596
- 75797-368
- 75798-598
- 76398-374
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1