Orders LinkContactUsLinkComponent
The Apprentice Doctor®Phlebotomy Practice Kit
The Apprentice Doctor®Phlebotomy Practice Kit
Phlebotomy practice kit, 1
Catalog # 470324-236
Supplier #:  AD603-INJ
The Apprentice Doctor®Phlebotomy Practice Kit
Catalog # 470324-236
Supplier:  
The Apprentice Doctor®Phlebotomy Practice Kit
The Apprentice Doctor®Phlebotomy Practice Kit
Phlebotomy practice kit, 1
Discontinued
Product Details & Documents
Practice on a real patient!
  • Wearable and position able.
  • Hard plastic back and sides To prevent needle pass through.
Teach students how to safely give injections and draw blood with this compact, wearable, phlebotomy trainer. When used on a simulator, it adds an element of positioning to the task. If used with a standard patient, it can be used as part of a standard exam protocol. The veins are large and comparatively shallow, and the trainer has a hard plastic case to prevent needle pass through. The textured surface prevents prior needlesticks from being obvious, and the latex tubing provides a longer life than many silicon models.
Caution: For educational and training use only. This product contains latex.
Specifications
  • Description:
    Phlebotomy practice kit
  • No. of lab groups:
    1