Radial Purge Extension Window for PerkinElmer Optima ICP-OES Instruments, PerkinElmer
Catalog #: CA97013-472
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Specifications
- Description:Radial purge extension window for Optima
- Cat. no.:CA97013-472
- Supplier No.:N0770944
Specifications
About this item
This radial view purge extension window is used with PerkinElmer Optima™ 5x00, 7100, 7200, 7300 DV, and 8300 ICP-OES series instruments.
The radial view purge extension window is a replaceable quartz window that slides into the mount between the torch and the optics for the radial view.
Customers Who Bought This Also Bought