MICROCHEM® AlphaTec® 2000 Ts Plus Coveralls, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
Superior breathable microporous laminate technology offering Type 4/5/6 protection.
- Proven protective barrier to low concentration liquid chemicals
- Moisture vapor permeable fabric ('breathable') to help reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Anti-static
- Ultra low linting, reduced risk of contamination in critical areas
- Silicone-free, critical in spray painting applications
- Tunnelled elasticated hood, wrists, and ankles
The MICROCHEM® by AlphaTec® 2000 Ts Plus coveralls provide both protection and comfort, with exceptional liquid and particulate protection. Ideal for a wide range of industrial applications.
Certifications: AATCC76; ASTM F1671. Tested according to EN 1149-5 and AATTC 76.
Specifications
- Catalog No:
- 76304-500
- 76304-494
- 76304-492
- 76304-498
- 76304-496
- 76304-486
- 76304-490
- 76304-488
- Fabric:
- Microporous polyethylene film
- Microporous polyethylene film
- Microporous polyethylene film
- Microporous polyethylene film
- Microporous polyethylene film
- Microporous polyethylene film
- Microporous polyethylene film
- Microporous polyethylene film
- Garment application:
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial