AlphaTec® 87-086 Natural Rubber Gloves, Ansell
Supplier: Ansell Canada
Certificates
About this item
Flocked, natural rubber gloves are comfortable and food compliant.
- Beaded cuff for tear resistance and easy donning
- Food approved
- Good abrasion resisstance
- 100% cotton flock lining
The AlphaTec® 87-086 natural rubber latex glove is flock lined and lightweight. It is resistant, flexible, and very comfortable. Ideally suited for use in low hazard areas.
Certifications: EN 420:2003 + A1:2009; FDA 21 CFR; CFIA.
Packaging: Gloves are packaged 12 pairs per bag, 12 bags per carton.
Specifications
- Catalog No:
- 75795-466
- 75795-468
- 75795-470
- 75795-472
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Liner material:
- Flock
- Flock
- Flock
- Flock
- Finish:
- Reversed lozenge
- Reversed lozenge
- Reversed lozenge
- Reversed lozenge
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
Specifications
Customers Who Bought This Also Bought