Shippers Declaration for Dangerous Goods Form, Labelmaster
Catalog #: 101378-106
Supplier: Label Master
Specifications
- Description:1-part blank laser form
- Height:11"
- Material:PP/cellulose
- Cat. no.:101378-106
- Width:8.5"
- Supplier No.:F07LB
Specifications
About this item
Comply with air shipment regulations with shippers declaration for dangerous goods form.
- Red and white striped border
- Available in multiple formats
This is required for all international shipments of dangerous goods/hazardous materials by air. The form includes air certification, an area for the 24-hour emergency response phone number, and the declaration statement for air eligibility. Use Masterform software to fill out the laser forms.
Customers Who Bought This Also Bought