Labels 'Inhalation Hazard', Labelmaster
Catalog #: 101376-382
Supplier: Label Master
Specifications
- Description:Preprinted labels
- Material:Semi-gloss paper
- Thickness:5.85 mil
- Cat. no.:101376-382
- Packaging:On roll
- Legend:Inhalation Hazard
- Dimensions:100×100 mm (3.94×3.94")
- Supplier No.:HML29
Specifications
About this item
Ship with confidence when using Worded Inhalation Hazard labels, revised to meet the design requirements of 49 CFR 172.407.
- Printed with light-fast inks for fade resistance
- Adhesives are formulated for use in various climates and environments
- Once affixed, labels won't fade or fall off
- Outstanding chemical and abrasion resistance
- White and black
In addition to complying with directives set by the DOT, these also meet the design and durability standards of 49 CFR, ICAO, and other international air regulations.
Customers Who Bought This Also Bought