Labels 'Hazardous Materials', Labelmaster
Catalog #: 101377-750
Supplier: Label Master
Specifications
- Description:Preprinted labels
- Material:Fluorescent red paper
- Thickness:7.1 mil
- Cat. no.:101377-750
- Packaging:On roll
- Core Size:3"
- Legend:Hazardous Materials
- Dimensions:76.2×76.2 mm (3×3")
- Supplier No.:LT208
Specifications
About this item
High-visibility warehouse labels instantly command attention wherever they're used.
- Strong adhesion to corrugated boxes
- Made of paper
- Bold lettering to assure important messaging is seen and read
When it has to be visible and has to be noticed, it has to be high-visibility warehouse labels. Hazardous materials labels alert shipping and receiving personnel to special handling instructions.
Customers Who Bought This Also Bought