Labels 'Dry Ice', Labelmaster
Catalog #: 101377-782
Supplier: Label Master
Specifications
- Description:Preprinted labels
- Material:Semi-gloss paper
- Thickness:5.85 mil
- Cat. no.:101377-782
- Packaging:On roll
- Core Size:3"
- Legend:Dry Ice UN1845
- Dimensions:152.4×152.4 mm (6×6")
- Supplier No.:HML11-DICE
Specifications
About this item
Labelmaster’s paper Dry Ice Labels have UN1845 Dry Ice preprinted on them.
- Meet the design and durability standards of 49 CFR, ICAO, and other international air regulations
- Labels will not fade or fall off
- Printed with light-fast inks for high durability
Labels feature blanks to fill in the dry ice quantity, the shipper's name, and the consignee. These Hazard Class 9 labels help maintain compliance with standards set forth by the DOT. Adhesives are formulated for use in various climates and environments. Hazard Class 9 paper labels have outstanding chemical and abrasion resistance, ensure strong adhesion to corrugated boxes, and help to keep users in regulatory compliance per the 49 CFR.
Customers Who Bought This Also Bought