KIMBLE® Micro DUALL® Tissue Grinder, All-Glass, DWK Life Sciences
Catalog #: KT885470-0000
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Capacity:200 μl
- Description:Micro tissue grinder
- Cat. no.:KT885470-0000
- Overall Length:33 mm
- Tube Dimensions:8 mm
- Pestle Length × Shaft O.D.:75×3 mm
- Supplier No.:885470-0000
Specifications
About this item
Designed for dispersing small quantities of tissue in buffer solution.
- PTFE bearing prevents misalignment and avoids aerosol formation
Suitable for muscle and heart tissues.
Customers Who Bought This Also Bought