KIMBLE® ULTRA-WARE® HPLC Reservoir Adapter, GL45, DWK Life Sciences
Catalog #: KT953905-0000
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- ST Joint:GL 45
- Description:HPLC reservoir adapter
- Material:PTFE
- Size:45 mm
- Cat. no.:KT953905-0000
- Inner ST Joint:40/35
- Supplier No.:953905-0000
Specifications
About this item
Designed to convert GL 45 threads to standard taper 40/35 inner joints.
- Autoclavable
- Allows filter membrane support glassware with 40/35 outer joints to be used on ULTRA-WARE®
- Highly chemically resistant PTFE remains nonbrittle at sub-zero temperatures
Customers Who Bought This Also Bought