D-(+)-Glucose, anhydrous, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP, SAFC®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: Corn sugar,Glucose,D-(+)-Dextrose,(2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal,Glucolin,Dextrose
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MW:180.16 g/mol
MDL Number:MFCD00148912
:Storage Temperature: Ambient
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 146 °C
:Flash Pt:
:Flash Pt:
:M.W. 180.16 g/mol
:D-(+)-Glucose
:146
Storage Temperature:Ambient
:12,04467
:14,04459
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (calc. on anhydrous substance):97.5 - 102.0 %
- Identity (IR):passes test
- Identity (spec. rotation calc. on anhydrous substance):passes test
- Identity (HPLC):passes test
- Identity (water):passes test
- Appearance of solution:passes test
- Color and clarity of solution:passes test
- Titrable acid:≤ 0.002 meq/g
- Conductivity:≤ 20 µS/cm
- Spec. rotation (α25/D):52.5 - 53.0 °
- In water insoluble matter:≤ 0.005 %
- Chloride (Cl):≤ 0.01 %
- Sulfate (SO₄):≤ 0.005 %
- Heavy metals (as Pb):≤ 5 ppm
- Fe (Iron):≤ 5 ppm
- Related substances [sum of impurities A (maltose) and B (isomaltose)!:≤ 0.4 %
- Related substances [Impurity C (maltotriose)!:≤ 0.2 %
- Related substances [Impurity D (fructose)!:≤ 0.15 %
- Related substances (unspecified impurities):≤ 0.10 %
- Related substances (Sum of all impurities):≤ 0.5 %
- Dextrin:passes test
- Soluble starch, sulfite:≤ 15 ppm
- Starch:passes test
- Residual solvents (ICH Q3C):excluded by the manufacturing process
- Loss on drying (105 °C):≤ 0.2 %
- Water (according to Karl Fischer):≤ 1.0 %
- Sulfated ash (600 °C):≤ 0.02 %
- Microbiological purity (Total aerobic microbial count (TAMC)):≤ 100 CFU/g
- Microbiological purity (Total yeast and mould count (TYMC)):≤ 100 CFU/g
- Bile tolerant Gram negative Bacteria (absent in 1 g):passes test
- Escherichia coli (absent in 1 g):passes test
- Pseudomonas aeruginosa (absent in 1 g):passes test
- Staphylococcus aureus (absent in 1 g):passes test
- Candida albicans (absent in 1 g):passes test
- Salmonella ssp (absent in 10 g):passes test
- Endotoxins:≤ 10 I.U./g
- Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).:
- Corresponds to Ph Eur,BP,USP,ACS:
- SAFC®, D-(+)-Glucose, anhydrous, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP:
Specification Test Results
Customers Who Bought This Also Bought