Axial Purge Extension Window for Optima ICP-OES Instruments, PerkinElmer
Catalog #: CA97013-474
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Specifications
- Description:Axial Purge Extension Window, DV Axial View
- Window Material:Quartz
- Cat. no.:CA97013-474
- Supplier No.:N0771116
Specifications
About this item
This axial view purge extension window is used with Optima™ 3x00 DV, 4x00 DV, 5x00 DV, 7x00 DV and 8x00 ICP-OES series instruments.
- The window is made from high-grade quartz
- Window should be replaced on a regular basis as part of a preventative maintenance schedule
- Maintain factory performance using genuine PerkinElmer replacement parts
The Axial View Purge Extension Window is a replaceable quartz window that slides into the purge extension between the torch and the optics of the instrument.
Customers Who Bought This Also Bought