KIMBLE® Stirrer Bearings, PTFE, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These solid stirrer bearings are ideal for use with Kontes series KT786500-0261, KT786500-0262, KT786500-0263, KT786500-0264 glass 10 mm precision ground stirring shafts and 19 mm precision ground glass stirring shafts.
Customers Who Bought This Also Bought