Avantor® Ultrasphere® ODS (C18), HPLC Columns
Supplier: HICHROM LIMITED
Certificates
About this item
Avantor® Ultrasphere® columns are available in variety of phases with 2 range of particle size.
- High reproducibility
- Widely referenced silica
- Optimal surface coverage for long column lifetime
- Narrow particle size distribution for high efficiency and improved resolution
Avantor® Ultrasphere® columns remain widely referenced within both industry and academia and are recognized to provide excellent chromatographic performance.
Specifications
- Catalog No:
- 76398-178
- 76398-180
- 76398-104
- 76398-082
- 76382-660
- 76398-084
- 76398-154
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes