Avantor® ACE® C4 Ultra-Inert, Analytical HPLC Columns, 3 µm
Supplier: HICHROM LIMITED
Certificates
About this item
Avantor® ACE® C4-300 columns provide great reproducibility and column lifetime. These columns are available in a wide range of particle sizes and dimensions from capillary to preparative. These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Ideal for a wide range of analytes differing in hydrophobicity
- Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
- Designed for peptide and protein separations
These columns are made of stainless steel.
Specifications
- Catalog No:
- 76382-592
- 76382-590
- Carbon load:
- 2.60%
- 2.60%
- Endcapped:
- Yes
- Yes
- Max. pressure:
- 400 bar (5802 psi)
- 400 bar (5802 psi)
- pH range:
- 2 - 8
- 2 - 8
- Surface area:
- 100 m²/g
- 100 m²/g
- USP code:
- L26
- L26
Specifications
Customers Who Bought This Also Bought