Avantor® ACE® Excel® SuperC18, Analytical HPLC/UHPLC Columns, 1.7 µm
Supplier: HICHROM LIMITED
Certificates
About this item
Avantor® ACE® Excel® SuperC18™ columns provide high stability across an extended pH range. These columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.
- Ultra-inert for maximum performance and reproducibility
- Ultra-low bleed for improved LC/MS compatibility
- Wide range of particle sizes (1.7 to 10 µm) and complementary phases
These columns are made of stainless steel.
Specifications
- Catalog No:
- 76394-452
- 76394-450
- 76394-454
- 76394-462
- 76394-460
- 76394-444
- 76394-448
- 76394-446
- 76382-652
- 76382-650
- Carbon load:
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- pH range:
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- Surface area:
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
Specifications
Customers Who Bought This Also Bought