KIMBLE® HI-VAC® Right-Angle Valves, with PTFE Plug and Extended Tip, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
This right-angle high-vacuum valve is designed with a precision external thread for use when fine control is needed.
- The accurately controlled valve seat and the extended tip on the PTFE plug allow control similar to a needle valve for both liquids and gases
- The FKM O-ring on the shaft backs up machined-in PTFE ring seals for high vacuum use which shield the O-ring from direct exposure to gases and liquids within the system
- Plug is interchangeable with other KONTES® valve bodies of the same size
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought